Ibotenic acid ≥95%, Moligand™
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
α-Amino-3-hydroxy-5-isoxazoleacetic acid, 2-Amino-2-(3-oxo-2,3-dihydroisoxazol-5-yl)acetic acid
A137136-10MG
A137136-1MG
A137136-25MG
A137136-50MG
A137136-5MG
ALA137136-10MGEA
351.41
USD
ALA137136-10MG
ALA137136-1MG
ALA137136-25MG
ALA137136-50MG
ALA137136-5MG
Ibotenic acid ≥95%, Moligand™
Ibotenic acid
Ibotenic acid has been used to perform surgery in adult male zebra finches (Taeniopygia guttata) to study reversing reinforcement-induced vocal changes.
Formula:
C₅H₆N₂O₄ MW: 158.113 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00069294 CAS Number: 2552-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...