2-Amino-5-fluorobenzothiazole ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Fluorobenzo[d]thiazol-2-amine

A137096-1G A137096-250MG A137096-5G
ALA137096-1GEA 93.01 USD
ALA137096-1G ALA137096250MG ALA137096-5G
2-Amino-5-fluorobenzothiazole ≥98%
2-Amino-5-fluorobenzothiazole

Application:2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.Remark:Stable under recommended storage conditions. Incompatible with oxidizing agents.

Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Melting Pt: 177 °C
MDL Number: MFCD05664560
CAS Number: 20358-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f31eacee88ac96b946c5f5776e05ef7ee6859fe&vid=6f31eacee88ac96b946c5f5776e05ef7ee6859fe&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR