6-Amino-1-naphthalenesulfonic acid ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-Amino-1-naphthalenesulphonic acid

A137050-100G A137050-25G A137050-5G
ALA137050-100GEA 253.56 USD
ALA137050-100G ALA137050-25G ALA137050-5G
6-Amino-1-naphthalenesulfonic acid ≥96%
6-Amino-1-naphthalenesulphonic acid

A product for proteomics research

Formula: C₁₀H₉NO₃S
MW: 223.2500 g/mol
Boiling Pt: 434.16 °C
Melting Pt: 173.38°C
Density: 1.5 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00035724
CAS Number: 81-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=060015d1fba41a3655c75212255deede251b0f5f&vid=060015d1fba41a3655c75212255deede251b0f5f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR