4-Amino-2-methylpyrimidine-5-carbonitrile ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A137005-10G A137005-5G
ALA137005-10GEA 127.21 USD
ALA137005-10G ALA137005-5G
4-Amino-2-methylpyrimidine-5-carbonitrile ≥95%
4-Amino-2-methylpyrimidine-5-carbonitrile
Formula: C₆H₆N₄
Storage Temperature: Refrigerator
MDL Number: MFCD00084875
CAS Number: 698-29-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3ce63bb3bd78af47cdd74709c89eb9a21fe3d55&vid=c3ce63bb3bd78af47cdd74709c89eb9a21fe3d55&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR