Alendronic acid ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 4-Amino-1-hydroxybutane-1,1-diphosphonic Acid, 4-Amino-1-hydroxybutane-1,1-diphosphonic acid, Alendronic Acid

A136993-1G A136993-5G
ALA136993-1GEA 16.06 USD
ALA136993-1G ALA136993-5G
Alendronic acid ≥98%, Moligand™
Alendronic acid

Alendronate acid is an inhibitor of FDPS. It is a bisphosphonate that inhibits osteoclast-mediated bone resorption. Unlike pyrophosphate, the endogenous regulator of bone turnover to which it is chemically related, alendronate specifically inhibits osteoclastic osteoclastic bone resorption, having no effect on mineralization of bone newly formed by osteoblasts.An inhibitor of FDPS

Formula: C₄H₁₃NO₇P₂
MW: 249.10 g/mol
Melting Pt: 235°C
MDL Number: MFCD00868112
CAS Number: 66376-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfbcde611d1f58618e48fb4c9d99f58c7122d226&vid=cfbcde611d1f58618e48fb4c9d99f58c7122d226&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR