2-(2-(2-azidoethoxy)ethoxy)ethanol ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Azido-PEG3-alcohol, N3-PEG3-OH, 1-Azido-8-hydroxy-3,6-dioxaoctane, 2-[2-(2-Azidoethoxy)ethoxy]ethanol, 8-Azido-3,6-dioxaoctanol
A136914-1G
A136914-250MG
A136914-5G
ALA136914-1GEA
112.01
USD
ALA136914-1G
ALA136914250MG
ALA136914-5G
2-(2-(2-azidoethoxy)ethoxy)ethanol ≥95%
2-(2-(2-azidoethoxy)ethoxy)ethanol
Formula:
C₆H₁₃N₃O₃ MW: 175.19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09753542 CAS Number: 86520-52-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...