(1H-Indol-5-yl)methanamine ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (1H-Indol-5-yl)methylamine, 5-(Aminomethyl)indole, (1H-Indol-5-yl)methanamine, Indole-5-methanamine

A136890-5G A136890-1G A136890-250MG A136890-25G
ALA136890-5GEA 322.91 USD
ALA136890-5G ALA136890-1G ALA136890250MG ALA136890-25G
(1H-Indol-5-yl)methanamine ≥95%
(1H-Indol-5-yl)methanamine

Reactant for preparation of: . HIV-1 integrase inhibitors targeting the catalytic domain and its ability for interaction with LEDGF/p75 . Inhibitors of Gli1-mediated transcription in the Hedgehog pathway . SCIO-469-like compounds for the inhibition of p38 MAP kinase . 4-(indol-5-ylamino)thieno[2,3-b]pyridine-5-carbonitriles as protein kinase c theta (PKC θ) inhibitors . Non-covalent thrombin inhibitors

Formula: C₉H₁₀N₂
MW: 146.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01719220
CAS Number: 81881-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c5fc0914a19c29edf5e4455236d76e1e7a41acf&vid=6c5fc0914a19c29edf5e4455236d76e1e7a41acf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR