5-Amino-4-cyano-1-phenyl-1H-pyrazole ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Amino-1-phenyl-1H-pyrazole-4-carbonitrile

A136652-1G A136652-250MG A136652-25G A136652-5G
ALA136652-1GEA 31.26 USD
ALA136652-1G ALA136652250MG ALA136652-25G ALA136652-5G
5-Amino-4-cyano-1-phenyl-1H-pyrazole ≥98%
5-Amino-4-cyano-1-phenyl-1H-pyrazole
Formula: C₁₀H₈N₄
MW: 184.20 g/mol
Melting Pt: 132-137°C
MDL Number: MFCD00020730
CAS Number: 5334-43-0
EINECS: 226-253-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41613b3ee74af17811c76a3fd907c34e02f48b3f&vid=41613b3ee74af17811c76a3fd907c34e02f48b3f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR