3-Acetoxybenzaldehyde ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Formylphenyl acetate

A136562-1G A136562-5G
ALA136562-1GEA 18.91 USD
ALA136562-1G ALA136562-5G
3-Acetoxybenzaldehyde ≥98% (by GC)
3-Acetoxybenzaldehyde
Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 138°C
Density: 1.17 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00016603
CAS Number: 34231-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=678cfc2cd18be605a351dece01921dc2d46142a5&vid=678cfc2cd18be605a351dece01921dc2d46142a5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR