4-Amino-3,5-dibromopyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A136486-100G A136486-1G A136486-25G A136486-5G
ALA136486-100GEA 1816.31 USD
ALA136486-100G ALA136486-1G ALA136486-25G ALA136486-5G
4-Amino-3,5-dibromopyridine ≥98% (by GC)
4-Amino-3,5-dibromopyridine
Formula: C₅H₄Br₂N₂
Melting Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00234048
CAS Number: 84539-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d5f73bb41812a0ff2af0691f5649977e7c8fe6b&vid=1d5f73bb41812a0ff2af0691f5649977e7c8fe6b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR