ɑ-Pyrone ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2H-Pyran-2-one, alpha-Pyrone

A136345-1G A136345-250MG A136345-50MG
ALA136345-1GEA 447.36 USD
ALA136345-1G ALA136345250MG ALA136345-50MG
ɑ-Pyrone ≥97% (by GC)
alpha-Pyrone
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 207 °C
Density: 1.2 g/cm³
Flash Pt: 94 °C
MDL Number: MFCD00006639
CAS Number: 504-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59b9464ff29adc4d12dd51af597048cd1e68c897&vid=59b9464ff29adc4d12dd51af597048cd1e68c897&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR