3-Amino-2-cyclohexen-1-one ≥98% (dry weight)

Supplier: ALADDIN SCIENTIFIC
Warning

A136183-1G A136183-25G A136183-5G
ALA136183-1GEA 10.36 USD
ALA136183-1G ALA136183-25G ALA136183-5G
3-Amino-2-cyclohexen-1-one ≥98% (dry weight)
3-Amino-2-cyclohexen-1-one
Formula: C₆H₉NO
Melting Pt: 132 °C
MDL Number: MFCD00013783
CAS Number: 5220-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0be479a86326b4f8bf8325054c47f642762a3402&vid=0be479a86326b4f8bf8325054c47f642762a3402&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR