2-Acetyl-5-cyanothiophene ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
5-Acetylthiophene-2-carbonitrile
A136066-1G
A136066-5G
ALA136066-1GEA
229.81
USD
ALA136066-1G
ALA136066-5G
2-Acetyl-5-cyanothiophene ≥97%
2-Acetyl-5-cyanothiophene
Formula:
C₇H₅NOS MW: 151.19 g/mol |
MDL Number:
MFCD00052314 CAS Number: 88653-55-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...