4-(Aminomethyl)piperidine ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A136022-1G
A136022-25G
A136022-5G
ALA136022-1GEA
9.41
USD
ALA136022-1G
ALA136022-25G
ALA136022-5G
4-(Aminomethyl)piperidine ≥98% (by GC)
4-(Aminomethyl)piperidine
Reactant: Evaulated as a small molecule inhibitor of CD4-gp120 binding Reagent Involved in solvothermal preparation of germanium oxide hydroxide 3D net zeotype Involved in zeolite synthesis Coupling agent for adhesive systems
Formula:
C₆H₁₄N₂ Boiling Pt: 200 °C Melting Pt: 25°C Flash Pt: 79°C Storage Temperature: Ambient |
MDL Number:
MFCD00006007 CAS Number: 7144-05-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...