Atropine ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

A135946-1G A135946-250MG A135946-5G
ALA135946-1GEA 18.91 USD
ALA135946-1G ALA135946250MG ALA135946-5G
Atropine ≥99%, Moligand™
Atropine
Formula: C₁₇H₂₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00022622
CAS Number: 51-55-8
EINECS: 200-104-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3d1f606329c82d255ad95d3e86123df893cbb1d&vid=c3d1f606329c82d255ad95d3e86123df893cbb1d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR