Morin ≥90% (by HPLC), Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2',3,4',5,7-Pentahydroxy-flavone, C.I. 75660, 2-(2,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2′,3,4′,5,7-Pentahydroxyflavone, Aluminum Ionophore I

A135920-1G A135920-250MG A135920-5G A135920-100MG
ALA135920-1GEA 209.86 USD
ALA135920-1G ALA135920250MG ALA135920-5G ALA135920100MG
Morin ≥90% (by HPLC), Moligand™
Morin

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate.

Formula: C₁₅H₁₀O₇
Storage Temperature: Freezer
MDL Number: MFCD00006826
CAS Number: 480-16-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af261889fee833f1d008da746553ce0990fe298b&vid=af261889fee833f1d008da746553ce0990fe298b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR