4-Pentylbenzoic acid ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amylbenzoic acid, 4-n-Pentylbenzoic acid, 4-Pentylbenzoic acid

A135814-100G A135814-25G A135814-5G
ALA135814-100GEA 104.41 USD
ALA135814-100G ALA135814-25G ALA135814-5G
4-Pentylbenzoic acid ≥98% (by titration)
4-Pentylbenzoic acid
Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Melting Pt: 89 °C
MDL Number: MFCD00002572
CAS Number: 26311-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a7c1022deb47c8033f40804fdadc0c2109cf0c8&vid=8a7c1022deb47c8033f40804fdadc0c2109cf0c8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR