4'-Acetoxyacetanilide ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
4-Acetamidophenyl acetate, Acetaminophen Acetate (Acetaminophen Impurity)
A135813-1G
A135813-5G
ALA135813-1GEA
9.41
USD
ALA135813-1G
ALA135813-5G
4'-Acetoxyacetanilide ≥98%
4'-Acetoxyacetanilide
An acetaminophen synthetic impurity.
Formula:
C₁₀H₁₁NO₃ MW: 193.1992 g/mol Melting Pt: 155 °C |
MDL Number:
MFCD00059205 CAS Number: 2623-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...