4'-Acetylbenzo-15-crown-5 (4'-acetylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) 95 GC_ASSAY_METHOD

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4'-Acetylbenzo-15-crown 5-Ether, 2,3-(4-Acetylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene

A135628-100MG A135628-1G A135628-250MG
ALA135628100MGEA 63.56 USD
ALA135628100MG ALA135628-1G ALA135628250MG
4'-Acetylbenzo-15-crown-5 (4'-acetylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) 95 GC_ASSAY_METHOD
4'-Acetylbenzo-15-crown-5 (4'-Acetylbenzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formula: C₁₆H₂₂O₆
Melting Pt: 95-99°C
MDL Number: MFCD00188564
CAS Number: 41757-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3df37912d507be83959e030e31d99fc9206e822&vid=e3df37912d507be83959e030e31d99fc9206e822&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR