Alosetron hydrochloride ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

A135530-100MG A135530-1G A135530-250MG
ALA135530100MGEA 263.06 USD
ALA135530100MG ALA135530-1G ALA135530250MG
Alosetron hydrochloride ≥98% (by HPLC)
Alosetron hydrochloride
Formula: C₁₇H₁₈N₄O·HCl
Melting Pt: 291 °C
Storage Temperature: Freezer
MDL Number: MFCD03453647
CAS Number: 122852-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d4bd536cdc535a0e1db50196a1bd9b1eb44204&vid=b2d4bd536cdc535a0e1db50196a1bd9b1eb44204&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR