5-Amino-1,2,3-thiadiazole ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1,2,3-Thiadiazol-5-amine, 5-Amino-1,2,3-Thiadiazole
A135511-100G
A135511-1G
A135511-25G
A135511-5G
ALA135511-100GEA
497.71
USD
ALA135511-100G
ALA135511-1G
ALA135511-25G
ALA135511-5G
5-Amino-1,2,3-thiadiazole ≥98% (by HPLC)
5-Amino-1,2,3-thiadiazole
Formula:
C₂H₃N₃S Melting Pt: 142 °C |
MDL Number:
MFCD00082810 CAS Number: 4100-41-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...