Methyl-3-amino-4-chlorobenzoate ≥98% (by HPLC)
Supplier: Aladdin Scientific
|
Warning
|
A135510-100G
A135510-10G
A135510-25G
A135510-5G
ALA135510-100GEA
555.66
USD
ALA135510-100G
ALA135510-10G
ALA135510-25G
ALA135510-5G
Methyl-3-amino-4-chlorobenzoate ≥98% (by HPLC)
Methyl 3-amino-4-chlorobenzoate
Formula:
C₈H₈ClNO₂ Melting Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD03470533 CAS Number: 40872-87-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...