1-Acetoxy-3-chloroacetone ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A135505-1G A135505-5G
ALA135505-1GEA 28.41 USD
ALA135505-1G ALA135505-5G
1-Acetoxy-3-chloroacetone ≥95% (by GC)
1-Acetoxy-3-chloroacetone
Formula: C₅H₇ClO₃
Density: 1.28 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00191365
CAS Number: 40235-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3265c0072fd36fe8fc3602ad38a69584abde5f1&vid=a3265c0072fd36fe8fc3602ad38a69584abde5f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR