2-Amino-5-phenylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Phenylpyridin-2-amine, 5-Phenyl-2-pyridylamine

A135460-100MG A135460-1G A135460-250MG A135460-5G
ALA135460100MGEA 27.46 USD
ALA135460100MG ALA135460-1G ALA135460250MG ALA135460-5G
2-Amino-5-phenylpyridine ≥97%
2-Amino-5-phenylpyridine
Formula: C₁₁H₁₀N₂
MW: 170.21 g/mol
Melting Pt: 135°C
MDL Number: MFCD01692452
CAS Number: 33421-40-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4bce5a85ff7a2e0ccbe3f3e338ad82e4e9689eb&vid=c4bce5a85ff7a2e0ccbe3f3e338ad82e4e9689eb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR