7-Amino-1H-indazole ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A135356-100G A135356-1G A135356-25G A135356-5G
ALA135356-100GEA 1579.76 USD
ALA135356-100G ALA135356-1G ALA135356-25G ALA135356-5G
7-Amino-1H-indazole ≥97%
7-Amino-1H-indazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 157-163°C
Storage Temperature: Ambient
MDL Number: MFCD00022790
CAS Number: 21443-96-9
EINECS: 244-391-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e287c3fa72194020c77cb06d2431205444385ae&vid=9e287c3fa72194020c77cb06d2431205444385ae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR