2-Amino-6-bromobenzonitrile ≥95%

Supplier: Aladdin Scientific
Warning

A135298-100G A135298-1G A135298-250MG A135298-25G A135298-5G
ALA135298-100GEA 485.36 USD
ALA135298-100G ALA135298-1G ALA135298250MG ALA135298-25G ALA135298-5G
2-Amino-6-bromobenzonitrile ≥95%
2-Amino-6-bromobenzonitrile
Formula: C₇H₅BrN₂
MDL Number: MFCD09834769
CAS Number: 77326-62-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59c8bb918a6313f85a8dca3ab27cecbe6afac8b0&vid=59c8bb918a6313f85a8dca3ab27cecbe6afac8b0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR