Methyl-1-aminocyclopropanecarboxylate hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Aminocyclopropane-1-carboxylic acid Methyl Ester Hydrochloride, 1-Aminocyclopropanecarboxylic acid methyll ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-Aminocyclopropanecarboxylic acid methyl ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate HCl

A135277-1G A135277-250MG A135277-5G
ALA135277-1GEA 85.41 USD
ALA135277-1G ALA135277250MG ALA135277-5G
Methyl-1-aminocyclopropanecarboxylate hydrochloride ≥95%
Methyl-1-aminocyclopropanecarboxylate hydrochloride
Formula: C₅H₁₀ClNO₂
MW: 151.5914 g/mol
Melting Pt: 186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078879
CAS Number: 72784-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f585b5e252ea23b7ec1ab9685e7852551b49f2e4&vid=f585b5e252ea23b7ec1ab9685e7852551b49f2e4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR