3-Methoxypropene ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Allyl methyl ether

A135232-1G A135232-25G A135232-5G
ALA135232-1GEA 17.01 USD
ALA135232-1G ALA135232-25G ALA135232-5G
3-Methoxypropene ≥97% (by GC)
3-Methoxypropene

Application:Allyl methyl ether it used as pharmaceutical intermediate.Â

Formula: C₄H₈O
Boiling Pt: 42-43 °C
Density: 0.768 g/cm³
Flash Pt: -23°C
Storage Temperature: Refrigerator
MDL Number: MFCD00008649
CAS Number: 627-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=996e88f8404e30ad98a501fd7d65c825f33359fc&vid=996e88f8404e30ad98a501fd7d65c825f33359fc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR