2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A135147-1G A135147-250MG A135147-10G
ALA135147-1GEA 25.56 USD
ALA135147-1G ALA135147250MG ALA135147-10G
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile ≥97%
2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
Formula: C₉H₁₀N₂S
Melting Pt: 141-144°C
MDL Number: MFCD00128278
CAS Number: 4651-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34bd7a441d8c1e67b7f037bfd80be83bbdaadad0&vid=34bd7a441d8c1e67b7f037bfd80be83bbdaadad0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR