2-Amino-5-bromo-3,4-dimethylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A135106-10G A135106-1G A135106-250MG A135106-25G A135106-5G
ALA135106-10GEA 434.06 USD
ALA135106-10G ALA135106-1G ALA135106250MG ALA135106-25G ALA135106-5G
2-Amino-5-bromo-3,4-dimethylpyridine ≥97%
2-Amino-5-bromo-3,4-dimethylpyridine

It is an active pharmaceutical ingredient.

Formula: C₇H₉BrN₂
MW: 201.07 g/mol
Melting Pt: 133-135°C
MDL Number: MFCD02093947
CAS Number: 374537-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0698b0a9e09c6263fe320d5fa7dd441a2758831f&vid=0698b0a9e09c6263fe320d5fa7dd441a2758831f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR