2-Amino-4'-methoxyacetophenone hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A134954-25G A134954-5G A134954-1G A134954-250MG
ALA134954-25GEA 784.61 USD
ALA134954-25G ALA134954-5G ALA134954-1G ALA134954-250MG
2-Amino-4'-methoxyacetophenone hydrochloride ≥95%
2-Amino-4'-methoxyacetophenone hydrochloride

2-Amino-4′-methoxyacetophenone hydrochloride is an organic building block.

Formula: C₉H₁₂ClNO₂
Melting Pt: 190-193°C
MDL Number: MFCD00193078
CAS Number: 3883-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06454c81f6fdf3e09f9314cd31eebb399039fbdd&vid=06454c81f6fdf3e09f9314cd31eebb399039fbdd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR