3-Amino-5-methylpyridine ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Amino-3-picoline

A134931-100G A134931-1G A134931-250MG A134931-25G A134931-5G
ALA134931-100GEA 1433.46 USD
ALA134931-100G ALA134931-1G ALA134931250MG ALA134931-25G ALA134931-5G
3-Amino-5-methylpyridine ≥97% (by GC)
3-Amino-5-methylpyridine
Formula: C6H8N2
MW: 108.1411 g/mol
Boiling Pt: 155°C
Melting Pt: 63.0 - 67.0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112508
CAS Number: 3430-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76e85e1fe3a62951b92678226b71f57d4cd5f1ac&vid=76e85e1fe3a62951b92678226b71f57d4cd5f1ac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR