WR-1065 dihydrochloride ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
2-[(3-Aminopropyl)amino]ethanethiol dihydrochloride, Amifostine Thiol Dihydrochloride
A134805-10MG
A134805-50MG
ALA134805-10MGEA
60.71
USD
ALA134805-10MG
ALA134805-50MG
WR-1065 dihydrochloride ≥95%
WR-1065 dihydrochloride
Amifostine Thiol Dihydrochloride is a metabolite of Amifostine.A metabolite of Amifostine
Formula:
C₅H₁₆Cl₂N₂S Storage Temperature: Freezer |
MDL Number:
MFCD00460339 CAS Number: 14653-77-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...