N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 7-Azagramine, 3-(Dimethylaminomethyl)-1H-pyrrolo[2,3-b]pyridine,7-Azagramine,Dimethyl-(1H-pyrrolo[2,3-b]-3-pyridinylmethyl)amine

A134688-1G A134688-250MG
ALA134688-1GEA 88.26 USD
ALA134688-1G ALA134688250MG
N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine ≥97%
N,N-Dimethyl-1-(1H-pyrrolo[2,3-b]pyridin-3-yl)methanamine
Formula: C₁₀H₁₃N₃
MW: 175.24 g/mol
Melting Pt: 144 - 149 °C
MDL Number: MFCD01580034
CAS Number: 5654-92-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f799720f113c2262c685e71c421fd69f2d916bdd&vid=f799720f113c2262c685e71c421fd69f2d916bdd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR