Methyl-α-bromoisobutyrate ≥99%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
2-Bromo-2-methylpropionic acid methyl ester, Methyl 2-Bromo-2-methylpropanoate, Methyl 2-bromo-2-methylpropionate, alpha-Bromoisobutyricacidmethylester, Methyl 2-bromo-2-methylpropanoate, Methyl α-bromoisobutyrate, Methyl 2-bromoisobutyrate
A134370-100ML
A134370-25ML
ALA134370100MLEA
40.76
USD
ALA134370100ML
ALA134370-25ML
Methyl-α-bromoisobutyrate ≥99%
Methyl-α-bromoisobutyrate
Methyl α-bromoisobutyrate was used in Reformatsky reaction to prepare 1,4-disubstituted 3,3-dimethylazetidin-2-ones.Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with a methyl end group
Formula:
C₅H₉BrO₂ MW: 181.029 g/mol Density: 1.399 g/cm³ Flash Pt: 49 °C |
MDL Number:
MFCD00017854 CAS Number: 23426-63-3 EINECS: 245-657-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...