Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Carbethoxycytosine, Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate, 4-Amino-2-hydroxy-pyrimidine-5-carboxylic acid ethyl ester

A134347-1G A134347-250MG A134347-50MG
ALA134347-1GEA 949.91 USD
ALA134347-1G ALA134347250MG ALA134347-50MG
Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate ≥95%
Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate
Formula: C₇H₉N₃O₃
MW: 183.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00552904
CAS Number: 20187-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf6070a0f8cbcb5a2123f1221d9364b8ece1d8fd&vid=cf6070a0f8cbcb5a2123f1221d9364b8ece1d8fd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR