2-Amino-6-bromobenzothiazole ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

A134312-1G A134312-25G A134312-5G
ALA134312-1GEA 13.21 USD
ALA134312-1G ALA134312-25G ALA134312-5G
2-Amino-6-bromobenzothiazole ≥97% (by titration)
2-Amino-6-bromobenzothiazole

Reaction of 4-bromoaniline with acetic acid and potassium thiocynate in solution of bromine in acetic acid yields 2-amino-6-bromobenzothiazole. 2-Amino-6-bromobenzothiazole can also be synthesized from 2-aminobenzothiazole via monobromination with silica-supported quinolinium tribromide.

Application:2-Amino-6-bromobenzothiazole may be used in the synthesis of the following2-amino-6-arylbenzothiazole compounds:6-p-tolylbenzo[d]thiazole-2-amine6-(4chlorophenyl)benzo[d]thiazole-2-amine6-(4-methoxyphenyl)benzo[d]thiazole-2-amine6-(3,5-bis(triflouromethyl)phenyl)benzo[d]thiazole-2-amine6-phenylbenzo[d]thiazole-2-amine

Formula: C₇H₅BrN₂S
Melting Pt: 214°C
Storage Temperature: Ambient
MDL Number: MFCD00152229
CAS Number: 15864-32-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9357e5a8cd8a3c89fe11fb17ef7ba6da09dfc77&vid=c9357e5a8cd8a3c89fe11fb17ef7ba6da09dfc77&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR