6-Morpholino-3-pyridinamine ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
6-Morpholinopyridin-3-amine, 5-Amino-2-(4-morpholinyl)pyridine
A134187-5G
A134187-1G
A134187-250MG
ALA134187-5GEA
970.81
USD
ALA134187-5G
ALA134187-1G
ALA134187250MG
6-Morpholino-3-pyridinamine ≥97%
6-Morpholino-3-pyridinamine
Application:5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.
Formula:
C₉H₁₃N₃O MW: 179.22 g/mol Melting Pt: 127-130°C |
MDL Number:
MFCD00119562 CAS Number: 52023-68-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...