5-Amino-4,6-dichloro-2-methylpyrimidine ≥97%

Supplier: Aladdin Scientific
Warning

A134148-1G A134148-5G
ALA134148-1GEA 13.21 USD
ALA134148-1G ALA134148-5G
5-Amino-4,6-dichloro-2-methylpyrimidine ≥97%
5-Amino-4,6-dichloro-2-methylpyrimidine
Formula: C₅H₅Cl₂N₃
Melting Pt: 70-73°C
Storage Temperature: Ambient
MDL Number: MFCD00194053
CAS Number: 39906-04-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=838782257b135a1eb179f7f0fdcdd2d68e6f2323&vid=838782257b135a1eb179f7f0fdcdd2d68e6f2323&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR