Carbazochrome ≥99% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A133989-25G
A133989-5G
A133989-100G
A133989-1G
ALA133989-25GEA
47.41
USD
ALA133989-25G
ALA133989-5G
ALA133989-100G
ALA133989-1G
Carbazochrome ≥99% (by HPLC)
Carbazochrome
Formula:
C₁₀H₁₂N₄O₃ Melting Pt: 214 °C |
MDL Number:
MFCD00059705 CAS Number: 69-81-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...