Astemizole ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-(4-Fluorobenzyl)-2-(1-[4-methoxyphenethyl]piperidin-4-yl)aminobenzimidazole, astemizole

A133983-10MG A133983-250MG A133983-50MG
ALA133983-10MGEA 65.46 USD
ALA133983-10MG ALA133983250MG ALA133983-50MG
Astemizole ≥98%, Moligand™
Astemizole
Formula: C₂₈H₃₁FN₄O
Storage Temperature: Refrigerator
MDL Number: MFCD00153919
CAS Number: 68844-77-9
EINECS: 272-441-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f51fed4128a7c970e2f2925c7f4164e5159a8a8a&vid=f51fed4128a7c970e2f2925c7f4164e5159a8a8a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR