α,α'-Dibromo-m-xylene ≥97% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Product Introduction:α,α′-Dibromo-m-xylene has been used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macrocyclic salt.
Formula:
C₈H₈Br₂ Boiling Pt: 140°C Melting Pt: 74-76°C |
MDL Number:
MFCD00000178 CAS Number: 626-15-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...