1-Acetyl-2-imidazolidinone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-Acetylimidazolidin-2-one

A133901-1G A133901-250MG A133901-5G
ALA133901-1GEA 11.31 USD
ALA133901-1G ALA133901250MG ALA133901-5G
1-Acetyl-2-imidazolidinone ≥98% (by GC)
1-Acetyl-2-imidazolidinone
Formula: C₅H₈N₂O₂
Melting Pt: 185 °C
MDL Number: MFCD00988254
CAS Number: 5391-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aaab639f8668de438174372b5ae7dd3cd09b538d&vid=aaab639f8668de438174372b5ae7dd3cd09b538d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR