4-Amino-2-bromobenzonitrile ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A133899-100G
A133899-1G
A133899-25G
A133899-5G
ALA133899-100GEA
2487.01
USD
ALA133899-100G
ALA133899-1G
ALA133899-25G
ALA133899-5G
4-Amino-2-bromobenzonitrile ≥95%
4-Amino-2-bromobenzonitrile
Formula:
C₇H₅BrN₂ |
MDL Number:
MFCD03695814 CAS Number: 53312-82-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...