3-Amino-2-naphthol ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A133883-5G A133883-1G A133883-250MG A133883-25G
ALA133883-5GEA 147.16 USD
ALA133883-5G ALA133883-1G ALA133883250MG ALA133883-25G
3-Amino-2-naphthol ≥96%
3-Amino-2-naphthol
Formula: C₁₀H₉NO
Melting Pt: 229-238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004113
CAS Number: 5417-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e113fabbad852988cfd92a13ad60ccf306ed028f&vid=e113fabbad852988cfd92a13ad60ccf306ed028f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR