Azomethine H hydrate

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]naphthalene-2,7-disulphonic acid hydrate, 8-Hydroxy-1-[[(2-hydroxyphenyl)methylene]amino]naphthalene-3,6-disulphonic acid hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid hydrate, Azomethine-H hydrate

A133643-100G A133643-10G A133643-1G A133643-25G A133643-5G
ALA133643-100GEA 1827.71 USD
ALA133643-100G ALA133643-10G ALA133643-1G ALA133643-25G ALA133643-5G
Azomethine H hydrate
Azomethine H hydrate

A highly sensitive reagent for testing boron

Formula: C₁₇H₁₃NO₈S₂·H₂O
MW: 441.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00066536
CAS Number: 32266-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9e5df188a1027b46d28787c964c69119721a942&vid=d9e5df188a1027b46d28787c964c69119721a942&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR