3-Methoxyanthranilic acid ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A133637-100G A133637-1G A133637-25G A133637-5G
ALA133637-100GEA 834.96 USD
ALA133637-100G ALA133637-1G ALA133637-25G ALA133637-5G
3-Methoxyanthranilic acid ≥98% (by HPLC)
3-Methoxyanthranilic acid
Formula: C₈H₉NO₃
Melting Pt: 172 °C
MDL Number: MFCD00075178
CAS Number: 3177-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ba492919ae6434f0ebf341847f89043ebc28a81&vid=9ba492919ae6434f0ebf341847f89043ebc28a81&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR