2-Amino-5-(trifluoromethoxy)benzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-amino-5-(trifluoromethoxy)benzoic acid

A133609-1G A133609-5G
ALA133609-1GEA 15.11 USD
ALA133609-1G ALA133609-5G
2-Amino-5-(trifluoromethoxy)benzoic acid ≥98%
2-Amino-5-(trifluoromethoxy)benzoic acid
Formula: C₈H₆F₃NO₃
MW: 221.14 g/mol
MDL Number: MFCD00176651
CAS Number: 83265-56-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b7546185b9940a8becfe2cbb3685ed4764421ba&vid=9b7546185b9940a8becfe2cbb3685ed4764421ba&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR