3-Amino-4-chlorobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A133477-100G A133477-25G A133477-5G
ALA133477-100GEA 1553.16 USD
ALA133477-100G ALA133477-25G ALA133477-5G
3-Amino-4-chlorobenzonitrile ≥98%
3-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
Melting Pt: 90 °C
MDL Number: MFCD00017089
CAS Number: 53312-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a29e3e62dace90e0853d24b5207ad4cdced252b&vid=9a29e3e62dace90e0853d24b5207ad4cdced252b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR