5-Amino-2-methylbenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Amino-2-methylbenzoic Acid, 2-Methyl-5-aminobenzoic acid, 5-Amino-o-toluic acid

A133352-100G A133352-1G A133352-25G A133352-5G
ALA133352-100GEA 448.31 USD
ALA133352-100G ALA133352-1G ALA133352-25G ALA133352-5G
5-Amino-2-methylbenzoic acid ≥97%
5-Amino-2-methylbenzoic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 197 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06208351
CAS Number: 2840-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR